Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin8 Antibody (mAbGPa) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395315 0.025 mg
NBP253121 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB395315 Supplier Novus Biologicals Supplier No. NBP2531210.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin8 Monoclonal specifically detects Actin8 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen Actin8
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone mAbGPa
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.02% Sodium Azide
Gene Alias ACT8, Actin-8
Host Species Mouse
Immunogen Purified Arabidopsis Actin8
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Signaling
Primary or Secondary Primary
Test Specificity This mouse monoclonal antibody was generated against plant actin ACT8 and recognizes all tested subclasses of plant actins (arabidopsis ACT1, 2, 3, 4, 7, 8, 11, 12) and Dictyosetelium actin.
Target Species Plant
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.