Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin8 Antibody (mAbGPa) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
| Antigen | Actin8 |
|---|---|
| Clone | mAbGPa |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
Actin8 Monoclonal specifically detects Actin8 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
| Actin8 | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Signaling | |
| PBS with 0.02% Sodium Azide | |
| Purified Arabidopsis Actin8 | |
| Primary | |
| This mouse monoclonal antibody was generated against plant actin ACT8 and recognizes all tested subclasses of plant actins (arabidopsis ACT1, 2, 3, 4, 7, 8, 11, 12) and Dictyosetelium actin. |
| mAbGPa | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Plant | |
| ACT8, Actin-8 | |
| IgG2b | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title