Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Active β2 Integrin (CD18) Mouse anti-Human, BUV 395, Clone: mAb 24 (also known as 24; m24; M24; mAb24), BD Horizon™
Mouse Monoclonal Antibody
$248.00 - $442.00
Specifications
Antigen | Active β2 Integrin (CD18) |
---|---|
Clone | mAb 24 (also known as 24; m24; M24; mAb24) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Description
Active β2 Integrin (CD18) Monoclonal antibody specifically detects Active β2 Integrin (CD18) in Human samples. It is validated for Flow CytometrySpecifications
Active β2 Integrin (CD18) | |
Flow Cytometry | |
Brilliant Ultraviolet 395 | |
Mouse | |
Human | |
ITGB2; Integrin beta-2; Integrin β2; LFA-1 β; LFA1b; beta 2 integrin; β2 integrin | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
mAb 24 (also known as 24; m24; M24; mAb24) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Fibronectin-purified Human Monocytes | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title