Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Active Caspase-3 Rabbit anti-Human, Mouse, BV650, Clone: C92-605.rMAb, BD Horizon™
Rabbit Monoclonal Antibody
$203.00 - $512.50
Specifications
Antigen | Active Caspase-3 |
---|---|
Clone | C92-605.rMAb |
Applications | Flow Cytometry, Control |
Classification | Monoclonal |
Conjugate | Brilliant Violet 650 |
Description
Active Caspase-3 Monoclonal antibody specifically detects Active Caspase-3 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
Active Caspase-3 | |
Flow Cytometry, Control | |
Brilliant Violet 650 | |
Rabbit | |
Human, Mouse | |
Apopain; CASP-3; CASP3; CC3; CPP-32; CPP32; Caspase 3; Yama | |
IgG κ | |
Affinity Chromatography |
C92-605.rMAb | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Active Caspase-3 Fragment | |
Primary | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title