Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-9 Rabbit anti-Human, Mouse, Clone: SC65-05, Novus Biologicals™
SDP

Catalog No. NBP267640 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267640 Supplier Novus Biologicals Supplier No. NBP267640

Rabbit Monoclonal Antibody

Caspase-9 Monoclonal specifically detects Caspase-9 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Caspase-9
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SC65-05
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10 - 1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Gene Symbols CASP9
Host Species Rabbit
Immunogen Recombinant protein within Human Caspase-9 aa 95-335/ 416. (SwissProt: P55211 Human; SwissProt: Q8C3Q9 Mouse)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 842
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.