Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin A Antibody (105), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB076289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076289 Supplier Novus Biologicals Supplier No. NBP290669AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Activin A Monoclonal specifically detects Activin A in Mouse samples. It is validated for ELISA.

Specifications

Antigen Activin A
Applications ELISA
Classification Monoclonal
Clone 105
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Activin A (NP_032406.1; Met1-Ser424).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3624
Test Specificity Has cross-reactivity in ELISA with: Human INHBA
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.