Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin C/Inhibin beta C Antibody (MM0076-4N17), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP212011R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212011R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212011R Supplier Novus Biologicals Supplier No. NBP212011R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.

Specifications

Antigen Activin C/Inhibin beta C
Applications Western Blot
Classification Monoclonal
Clone MM0076-4N17
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias inhibin beta C chain, IHBC, inhibin, beta C
Gene Symbols INHBC
Host Species Mouse
Immunogen Human recombinant Activin C/Inhibin beta C
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3626
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.