Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin RIB/ALK-4 Antibody (544310), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349169 Supplier Novus Biologicals Supplier No. FAB2221MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Activin RIB/ALK-4 Monoclonal specifically detects Activin RIB/ALK-4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Activin RIB/ALK-4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 544310
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ACTRIB, ACVRLK4ACTR-IB, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Activin RIB/ALK-4, Ser24-Glu126, Accession # P36896
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 91
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.