Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin RIIA Antibody (OTI5D1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270079R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270079R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270079R Supplier Novus Biologicals Supplier No. NBP270079R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Activin RIIA Monoclonal specifically detects Activin RIIA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Activin RIIA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5D1
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ACTRII, ACTRIIA, ACVR2ACTR-IIA, EC 2.7.11, EC 2.7.11.30
Gene Symbols ACVR2A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 214-513 of human Activin RIIA (NP_001607) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 92.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.