Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin RIIB Antibody (307), HRP
SDP

Catalog No. NB073858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073858 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073858 Supplier Novus Biologicals Supplier No. NBP290465H

Rabbit Monoclonal Antibody

Activin RIIB Monoclonal specifically detects Activin RIIB in Mouse samples. It is validated for ELISA.

Specifications

Antigen Activin RIIB
Applications ELISA
Classification Monoclonal
Clone 307
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ACTRIIB, ACTR-IIB, EC 2.7.11, EC 2.7.11.30, MGC116908
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Activin RIIB extracellular domain (NP_031423.1; Met 1-Thr 134).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 93
Test Specificity No cross-reactivity in ELISA with: Human ACVR2B
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.