Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTL6B Antibody, Novus Biologicals™
SDP

Catalog No. NBP215269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215269 Supplier Novus Biologicals Supplier No. NBP215269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACTL6B Polyclonal specifically detects ACTL6B in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ACTL6B
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.62mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O94805
Gene Alias 53 kDa BRG1-associated factor B, actin-like 6, actin-like 6B, Actin-related protein Baf53b, ACTL6, ArpNalpha, BAF53Bactin-like protein 6B, BRG1-associated factor 53B, hArpN alpha
Gene Symbols ACTL6B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BAF53B. The exact sequence is proprietary.
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 51412
Test Specificity Human. Expected to cross react based on sequence identity: Rhesus Monkey (100%).
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.