Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15688620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15688620 20 μL
NBP156886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15688620 Supplier Novus Biologicals Supplier No. NBP15688620UL

Rabbit Polyclonal Antibody

ACTR10 Polyclonal specifically detects ACTR10 in Human samples. It is validated for Western Blot.

Specifications

Antigen ACTR10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NZ32
Gene Alias actin-related protein 10 homolog (S. cerevisiae), Actin-related protein 11, ACTR11actin-related protein 10, Arp11, HARP11
Gene Symbols ACTR10
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ACTR10(actin-related protein 10 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of ACTR10 (NP_060947). Peptide sequence SLIQCPIDTRKQLAENLVVIGGTSMLPGFLHRLLAEIRYLVEKPKYKKAL.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55860
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.