Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR1A Antibody, Novus Biologicals™
SDP

Catalog No. NBP15307520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15307520 20 μL
NBP153075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15307520 Supplier Novus Biologicals Supplier No. NBP15307520UL

Rabbit Polyclonal Antibody

ACTR1A Polyclonal specifically detects ACTR1A in Human samples. It is validated for Western Blot.

Specifications

Antigen ACTR1A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P61163
Gene Alias Actin-RPV, alpha-centractin, ARP1 (actin-related protein 1, yeast) homolog A (centractin alpha), ARP1 actin-related protein 1 homolog A, centractin alpha (yeast), ARP1actin-RPV, centractin, Centrosome-associated actin homolog, CTRN1, FLJ52695, FLJ52800, FLJ55002
Gene Symbols ACTR1A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ACTR1A(ARP1 actin-related protein 1 homolog A, centractin alpha (yeast)) The peptide sequence was selected from the middle region of ACTR1A. Peptide sequence AIKERACYLSINPQKDETLETEKAQYYLPDGSTIEIGPSRFRAPE
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10121
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.