Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR2 Rabbit anti-Human, Mouse, Rat, Clone: 6B1I3, Novus Biologicals™
SDP

Catalog No. NB168519 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168519 100 μg
NB168520 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168519 Supplier Novus Biologicals Supplier No. NBP316714100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ACTR2 Monoclonal antibody specifically detects ACTR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ACTR2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 6B1I3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias actin-related protein 2, ARP2 (actin-related protein 2, yeast) homolog, ARP2 actin-related protein 2 homolog (yeast), ARP2Actin-like protein 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 295-394 of human ACTR2 (P61160).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10097
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.