Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR3 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB112264 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112264 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112264 Supplier Novus Biologicals Supplier No. NBP297009FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACTR3 Polyclonal specifically detects ACTR3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ACTR3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ACTR3 (NP_005712.1; Ala2-Ser418).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10096
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.