Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR3C Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB351637 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351637 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351637 Supplier Novus Biologicals Supplier No. NBP306502PCP

Rabbit Polyclonal Antibody

ACTR3C Polyclonal antibody specifically detects ACTR3C in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ACTR3C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Actin-Related Arp11, Actin-related protein 11, Arp11, ARP3 Actin-Related Protein 3 Homolog C (Yeast)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ACTR3C fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653857
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.