Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACVR2A Monoclonal Antibody (C11)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | ACVR2A |
---|---|
Clone | C11 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541541
|
Invitrogen™
MA541541 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543988
|
Invitrogen™
MA543988 |
20 μL |
Each for $243.00
|
|
|||||
Description
ACVR2A Monoclonal Antibody for Western Blot, IHC (P)
Activin A receptor kinase (ACVR2A) is a type II member of the TGF-beta family of receptor Serine/Threonine kinases. ACVR2A is involved in the activin and myostatin signaling pathway. (ACVR2A Aliases: ACVR2, ACTRII). These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling, and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Type II receptors are considered to be constitutively active kinases.Specifications
ACVR2A | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P27037 | |
92 | |
Recombinant protein Activin A Receptor Type II A. The antigen corresponds to amino acid range 20-135 of the target protein. | |
Antibody |
C11 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
ACVR2A | |
activin A receptor type 2A; activin A receptor type IIA; activin A receptor, type IIA; activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; Actrii; ActrIIa; ACTR-IIA; ACVR2; Acvr2a; rActR-II; TactrII; type II activin receptor | |
ACVR2A | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title