Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACY3 Antibody (OTI2D5), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N272329PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272329PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272329PEC5 Supplier Novus Biologicals Supplier No. NBP272329PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACY3 Monoclonal specifically detects ACY3 in Human, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen ACY3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2D5
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ACY-3aspartoacylase-2, Acylase III, Aminoacylase-3, aspartoacylase (aminocyclase) 3, EC 3.5.1.-, EC 3.5.1.15, HCBP1ASPA2, Hepatitis C virus core-binding protein 1, MGC9740
Gene Symbols ACY3
Host Species Mouse
Immunogen Full length recombinant protein of human ACY3 (NP_542389) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 91703.0
Target Species Human, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.