Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACYP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB090703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090703 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090703 Supplier Novus Biologicals Supplier No. NBP300018H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACYP1 Polyclonal specifically detects ACYP1 in Human samples. It is validated for ELISA.

Specifications

Antigen ACYP1
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias acylphosphatase 1, erythrocyte (common) type, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1, ACYPEacylphosphatase-1, EC 3.6.1.7
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ACYP1 (P07311; Ala 2-Lys 99).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 97
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.