Learn More
Liofilchem™ AD Fosfomycin
Shop All Liofilchem Products
Click to view available options
No. per Pack:
1 Ea.
6/Pk.
Description
- 1 panel of AD Fosfomycin 0.25-256
- 2 Reading Templates (transparent and black backgrounds)
- 1 Instructions Sheet also available at www.liofilchem.com/ifu-sds
Specifications
Specifications
Antimicrobial Agents | Fosfomycin |
Concentration | 0.25 to 256 μg/mL |
Product Type | Panel |
Sterility | Non-sterile |
Test Type | Media Testing |
No. per Pack | 1 Ea. |