Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM10 Antibody (163003), FITC, Novus Biologicals™
SDP

Catalog No. NB297136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297136 Supplier Novus Biologicals Supplier No. FAB1427F

Mouse Monoclonal Antibody

ADAM10 Monoclonal antibody specifically detects ADAM10 in Human samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF

Specifications

Antigen ADAM10
Applications Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 163003
Conjugate FITC
Dilution Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ADAM10, Thr214-Glu672, Accession # O14672
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 102
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.