Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM10 Antibody (RM0146-7H12), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP212015X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212015X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212015X Supplier Novus Biologicals Supplier No. NBP212015AF488

Rat Monoclonal Antibody

ADAM10 Monoclonal specifically detects ADAM10 in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen ADAM10
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone RM0146-7H12
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunofluorescence
Gene Alias a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
Gene Symbols ADAM10
Host Species Rat
Immunogen Mouse recombinant ADAM10
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 102
Test Specificity Detects mouse ADAM10 and cross reacts with human ADAM10. Weak cross-reactivity is observed to ADAM9 and ADAM15 in Western blotting.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.