Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM10 Mouse anti-Human, Clone: 6D39, Novus Biologicals™
SDP

Catalog No. NB019662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019662 0.02 mg
NBP23606805 0.05 mg
NBP236068 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB019662 Supplier Novus Biologicals Supplier No. NBP2360680.02MG

Mouse Monoclonal Antibody

ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ADAM10
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6D39
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with No Preservative
Gene Alias a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
Gene Symbols ADAM10
Host Species Mouse
Immunogen recombinant human ADAM-10 (extracellular domain)
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 102
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.