Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM15 Antibody (007), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB096818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096818 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096818 Supplier Novus Biologicals Supplier No. NBP289546C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ADAM15 Monoclonal specifically detects ADAM15 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.

Specifications

Antigen ADAM15
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ADAM15 (NP_997074.1; Met 1-Thr 696).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8751
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.