Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM15 Antibody (23G9), FITC, Novus Biologicals™
SDP

Catalog No. NB295476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295476 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295476 Supplier Novus Biologicals Supplier No. FAB935F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADAM15 Monoclonal antibody specifically detects ADAM15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen ADAM15
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 23G9
Conjugate FITC
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin
Host Species Mouse
Immunogen COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant human ADAM15, Asp207-Thr696, Accession # Q13444
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8751
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.