Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM23 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB052080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052080 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052080 Supplier Novus Biologicals Supplier No. NBP298071AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAM23 Polyclonal specifically detects ADAM23 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ADAM23
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias a disintegrin and metalloproteinase domain 23, ADAM metallopeptidase domain 23, disintegrin and metalloproteinase domain-containing protein 23, MDC-3, MDC3ADAM 23, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ADAM23 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8745
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.