Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM28 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25599525UL
Description
ADAM28 Polyclonal specifically detects ADAM28 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ADAM28 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L | |
Rabbit | |
Affinity Purified | |
RUO | |
10863 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ADAM28 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ELPGVKKYEVVYPIRLHPLHKREAKEPEQQEQFETELKYKMTINGKIAVLYLKKNKNLLAPGYTETYYNSTGKEITTSPQIMDDCYY | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°CC long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction