Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM29 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342757 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342757 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342757 Supplier Novus Biologicals Supplier No. NBP281697MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAM29 Polyclonal antibody specifically detects ADAM29 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ADAM29
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ADAM 29, ADAM metallopeptidase domain 29, Cancer/testis antigen 73, CT73a disintegrin and metalloproteinase domain 29, disintegrin and metalloproteinase domain-containing protein 29, EC 3.4.24, EC 3.4.24.58, svph1
Host Species Rabbit
Immunogen ADAM29 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ADAM29. The immunogen is located within amino acids 710 - 760 of ADAM29. Amino Acid Squence: EEEKIQRRPHELPPQSQPW
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11086
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.