Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM33 Antibody (508101), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB296086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296086 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296086 Supplier Novus Biologicals Supplier No. FAB5565R

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ADAM33 Monoclonal antibody specifically detects ADAM33 in Human samples. It is validated for Immunoprecipitation, Flow Cytometry, CyTOF

Specifications

Antigen ADAM33
Applications Immunoprecipitation, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 508101
Conjugate Alexa Fluor 647
Dilution Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAM33, Val30-Leu501 (predicted), Accession # Q9BZ11
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80332
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.