Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM8 Antibody (MM0079-6J12), HRP, Novus Biologicals™
SDP

Catalog No. NBP212020H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212020H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212020H Supplier Novus Biologicals Supplier No. NBP212020H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot.

Specifications

Antigen ADAM8
Applications Western Blot
Classification Monoclonal
Clone MM0079-6J12
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8
Gene Symbols ADAM8
Host Species Mouse
Immunogen Human recombinant ADAM8 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 101
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.