Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM9 Antibody (MM0080-8A15), PE, Novus Biologicals™
SDP

Catalog No. NB361461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361461 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361461 Supplier Novus Biologicals Supplier No. NBP212021PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADAM9 Monoclonal antibody specifically detects ADAM9 in Human samples. It is validated for Western Blot

Specifications

Antigen ADAM9
Applications Western Blot
Classification Monoclonal
Clone MM0080-8A15
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase
Host Species Mouse
Immunogen Human recombinant ADAM9 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8754
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.