Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS1 Antibody (OTI4H2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP271474A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271474A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271474A32 Supplier Novus Biologicals Supplier No. NBP271474AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ADAMTS1
Applications Western Blot
Classification Monoclonal
Clone OTI4H2
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH-1
Gene Symbols ADAMTS1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 412-644 of human ADAMTS1 (NP_008919) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9510
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.