Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS13 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291899 Supplier Novus Biologicals Supplier No. NB10055261F
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ADAMTS13 Polyclonal specifically detects ADAMTS13 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen ADAMTS13
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTPADAM-TS 13, vWF-cleaving protease, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease
Host Species Goat
Immunogen Maps to a region between residue 825 and 875 of human ADAM (a disintegrin-like and metalloprotease) metallopeptidase with thrombospondin type 1 motif, 13 using the numbering given in entry NP_620594.1 (GeneID 1109
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11093
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.