Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS14 Antibody, Unconjugated, Novus Biologicals™
SDP

Catalog No. NB300998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300998 Supplier Novus Biologicals Supplier No. NBP326063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAMTS14 Polyclonal antibody specifically detects ADAMTS14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen ADAMTS14
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Paraffin 1:200
Formulation PBS, 50% Glycerol
Gene Alias ADAM metallopeptidase with thrombospondin type 1 motif, 14,14
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADAMTS14, corresponding to a region within C-terminal amino acids.
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140766
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.