Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS15 Antibody (561819), HRP, Novus Biologicals™
SDP

Catalog No. NB368469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368469 Supplier Novus Biologicals Supplier No. FAB5149H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADAMTS15 Monoclonal antibody specifically detects ADAMTS15 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen ADAMTS15
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 561819
Conjugate HRP
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias A disintegrin and metalloproteinase with thrombospondin motifs 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, preproprotein, ADAM metallopeptidase with thrombospondin type 1 motif, 15, ADAM-TS 15, ADAM-TS15, ADAMTS-15, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, metalloprotease disintegrin 15 with thrombospondin domains, MGC126403
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS15, Gly18-Cys682, Accession # Q8TE58
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170689
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.