Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139886 Supplier Invitrogen™ Supplier No. PA5139886
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GLSGMIRTEE ADYFLRPLPS HLSWKLGRAA QGSSPSHVLY KRSTEPHAPG ASEVLVTSRT WELAHQPLHS Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 63%.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has high sequence similarity to the protein encoded by ADAMTS18, another family member.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS16
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAMTS16
Gene Accession No. Q8TE57
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 16; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 16; a disintegrin-like and metallopetidase (reprolysin type) with thrombospondin type 1 motif, 16; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 16; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 16 preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 16; ADAM metallopeptidase with thrombospondin type 1 motif, 16; ADAMs; ADAM-TS 16; ADAMTS16; ADAM-TS16; ADAMTS-16; ADAMTS16s; KIAA2029; metalloendopeptidases
Gene Symbols ADAMTS16
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ADAMTS16 (Product #RP-109138)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170690
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.