Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTSL-1/Punctin Mouse anti-Human, Clone: 1B2-8-2, Novus Biologicals™
SDP

Catalog No. NB119890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119890 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119890 Supplier Novus Biologicals Supplier No. NBP311265

Mouse Monoclonal Antibody

ADAMTSL-1/Punctin Monoclonal specifically detects ADAMTSL-1/Punctin in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen ADAMTSL-1/Punctin
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 1B2-8-2
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias ADAM-TS related protein 1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1
Host Species Mouse
Immunogen Monoclonal antibody 1B2-8-2 reacts with Punctin-1 (ADAMTSL-1).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92949
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.