Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTSL-1/Punctin Mouse anti-Human, Clone: 9D18, Novus Biologicals™
SDP

Catalog No. NBP236074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236074 0.1 mg
NB019663 0.02 mg
NBP23607405 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP236074 Supplier Novus Biologicals Supplier No. NBP236074

Mouse Monoclonal Antibody

ADAMTSL-1/Punctin Monoclonal specifically detects ADAMTSL-1/Punctin in Human samples. It is validated for Western Blot.

Specifications

Antigen ADAMTSL-1/Punctin
Applications Western Blot
Classification Monoclonal
Clone 9D18
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000
Gene Alias ADAM-TS related protein 1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1
Gene Symbols ADAMTSL1
Host Species Mouse
Immunogen recombinant human ADAMTSL-1
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92949
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.