Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTSL-1/Punctin Mouse anti-Human, Clone: 9D18, Novus Biologicals™

Mouse Monoclonal Antibody
$244.00 - $646.00
Specifications
| Antigen | ADAMTSL-1/Punctin |
|---|---|
| Clone | 9D18 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019663
![]() |
Novus Biologicals
NBP2360740.02MG |
0.02 mg |
Each for $244.00
|
|
|||||
NBP23607405
![]() |
Novus Biologicals
NBP2360740.05MG |
0.05 mg |
Each for $464.00
|
|
|||||
NBP236074
![]() |
Novus Biologicals
NBP236074 |
0.1 mg |
Each for $646.00
|
|
|||||
Description
ADAMTSL-1/Punctin Monoclonal specifically detects ADAMTSL-1/Punctin in Human samples. It is validated for Western Blot.Specifications
| ADAMTSL-1/Punctin | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Human | |
| 92949 | |
| recombinant human ADAMTSL-1 | |
| Primary |
| 9D18 | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Mouse | |
| ADAM-TS related protein 1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1 | |
| ADAMTSL1 | |
| IgG2 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title