Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTSL3 Antibody, Novus Biologicals™
SDP

Catalog No. NB396070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396070 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396070 Supplier Novus Biologicals Supplier No. NBP24862725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAMTSL3 Polyclonal antibody specifically detects ADAMTSL3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ADAMTSL3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias ADAMTSL-3, ADAMTS-like 3, ADAMTS-like protein 3, KIAA1233a disintegrin-like and metalloprotease domain with thrombospondin type Imotifs-like 3, MGC150716, MGC150717, punctin-2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: LSGNVSLLFNGSLLLQNVSLENEGTYVCIATNALGKAVATSVLHLLERRWPESRIVFLQGHKKYILQATNTRTNSNDPTGEPPPQEPFWEPGNWSHCSATCGH
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57188
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.