Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTSL5 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241120 0.1 mg
NB396497 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241120 Supplier Novus Biologicals Supplier No. NBP241120
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

ADAMTSL5 Polyclonal specifically detects ADAMTSL5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ADAMTSL5
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias ADAMTSL-5, ADAMTS-like 5, ADAMTS-like protein 5, thrombospondin type-1 domain-containing protein 6, thrombospondin, type I, domain containing 6, THSD6
Gene Symbols ADAMTSL5
Host Species Chicken
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ADAMTSL5. The immunogen is located within the last 50 amino acids of ADAMTSL5. Amino Acid Squence: YARPWSPAEDSRIRLTA
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339366
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.