Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTSL5 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | ADAMTSL5 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ADAMTSL5 Polyclonal specifically detects ADAMTSL5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ADAMTSL5 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
339366 | |
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ADAMTSL5. The immunogen is located within the last 50 amino acids of ADAMTSL5. Amino Acid Squence: YARPWSPAEDSRIRLTA | |
Primary |
1 mg/mL | |
Polyclonal | |
Chicken | |
Human, Mouse, Rat | |
ADAMTSL-5, ADAMTS-like 5, ADAMTS-like protein 5, thrombospondin type-1 domain-containing protein 6, thrombospondin, type I, domain containing 6, THSD6 | |
ADAMTSL5 | |
IgY | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title