Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Adapter for Conical Tubes
Shop All Eppendorf North America Products
Click to view available options
For Use With (Equipment):
Rotor A-4-81 and Rotor FA-45-6-30
Rotor FA-45-6-30
Description
- Available in packs of two or seven
- For use with Eppendorf Rotors
Specifications
Specifications
| Type | Adapter |
| Tube Type | Conical |
| Capacity | 15 mL |
| For Use With (Equipment) | Rotor A-4-81 and Rotor FA-45-6-30 |
| Color | Orange |
| No. per Pack | Set of 7 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction