Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAR Sheep anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151725 25 μg
NB151726 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151725 Supplier Novus Biologicals Supplier No. NBP31188625UG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

ADAR Polyclonal antibody specifically detects ADAR in Rat samples. It is validated for Western Blot

Specifications

Antigen ADAR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS
Gene Alias ADAR1DSH, adenosine deaminase, RNA-specific, DRADAP136, DSRADadenosine deaminase acting on RNA 1-A, EC 3.5.4, EC 3.5.4.-, G1P1double-stranded RNA-specific adenosine deaminase, IFI-4, IFI4dsRNA adenosine deaminase, interferon-induced protein 4, Interferon-inducible protein 4, K88DSRBP, p136,136 kDa double-stranded RNA-binding protein
Host Species Sheep
Immunogen Sheep were immunized with a GST fusion protein containing amino acid 1116-1176 of the rat DRADA coding sequence. The fusion protein was constructed in the pGEX-KG vector by inserting a fragment corresponding to nucleotides +3346 through +3525 of DRADA at the XbaI and XhoI sites in the vector. (Uniprot: P55266)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.