Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAR Sheep anti-Rat, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody
$280.00 - $487.50
Specifications
Antigen | ADAR |
---|---|
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ADAR Polyclonal antibody specifically detects ADAR in Rat samples. It is validated for Western BlotSpecifications
ADAR | |
Western Blot | |
Unconjugated | |
Sheep | |
Rat | |
ADAR1DSH, adenosine deaminase, RNA-specific, DRADAP136, DSRADadenosine deaminase acting on RNA 1-A, EC 3.5.4, EC 3.5.4.-, G1P1double-stranded RNA-specific adenosine deaminase, IFI-4, IFI4dsRNA adenosine deaminase, interferon-induced protein 4, Interferon-inducible protein 4, K88DSRBP, p136,136 kDa double-stranded RNA-binding protein | |
Sheep were immunized with a GST fusion protein containing amino acid 1116-1176 of the rat DRADA coding sequence. The fusion protein was constructed in the pGEX-KG vector by inserting a fragment corresponding to nucleotides +3346 through +3525 of DRADA at the XbaI and XhoI sites in the vector. (Uniprot: P55266) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
103 | |
IgG | |
Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title