Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAR1 Rat anti-Mouse, Human, Clone: RD4B11, MilliporeSigma™
Rat Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | ADAR1 |
---|---|
Clone | RD4B11 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE1790-25
|
MilliporeSigma
MABE179025UG |
25 μg |
Each for $192.35
|
|
|||||
MABE1790-100
|
MilliporeSigma
MABE1790100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-ADAR1, clone RD4B11 is a highly specific rat monoclonal antibody that targets ADAR1 and has been tested for use in Western Blotting.Specifications
ADAR1 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Human | |
Adar | |
IgG2a κ | |
Protein G Purified | |
Clone RD4B11 is a rat monoclonal antibody that detects Double-stranded RNA-specific adenosine deaminase in human and murine cells. |
RD4B11 | |
Western Blot | |
Unconjugated | |
Rat | |
Epigenetics and Nuclear Function | |
NP_062629.3 | |
His-tagged recombinant fragment corresponding to 401 amino acids from the internal region of murine Double-stranded RNA-specific adenosine deaminase (ADAR1). Immunogen sequnece is conserved isoforms 1 and 2. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title