Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Adavosertib
GREENER_CHOICE

Catalog No. p-7239453
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective Wee1 inhibitor

Adavosertib is a potent and selective Wee1 inhibitor (IC50 = 5.2 nM). In a panel of 223 serine/threonine or tyrosine kinases, Adavosertib is highly selective for Wee 1 over all except 8 kinases. In particular Adavosertib shows >100-fold selectivity over human Myt1. Adavosertib blocks CDC2Y15 phosphorylation and Gemcitabine (Cat. No. 3259)-induced DNA damage checkpoint, leading to premature mitotic entry and apoptosis in p53-deficient tumor cell lines. Adavosertib also potentiates the cytotoxic effects of Pemetrexed (Cat. No. 6185), Doxorubicin (Cat. No. 2252), Camptothecin (Cat. No. 1100) and Mitomycin C (Cat. No. 3258) in vitro. Adavosertib synergizes with gemcitabine to achieve tumor regressions, selectively in p53-deficient pancreatic cancer xenografts.

Specifications

Chemical Name or Material 1,2-Dihydro-1-[6-(1-hydroxy-1-methylethyl)-2-pyridinyl]-6-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-2-(2-propen-1-yl)-3 H-pyrazolo[3,4-d]pyrimidin-3-one
CAS 955365-80-7
Quantity 10 mg
Target Checkpoint Control Kinase Inhibitors
Molecular Formula C27H32N8O2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.