Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADCY6 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | ADCY6 |
---|---|
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118931
|
Invitrogen™
PA5118931 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122273
|
Invitrogen™
PA5122273 |
20 μL |
Each for $209.50
|
|
|||||
Description
ADCY6 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the adenylyl cyclase family of proteins, which are required for the synthesis of cyclic AMP. All members of this family have an intracellular N-terminus, a tandem repeat of six transmembrane domains separated by a cytoplasmic loop, and a C-terminal cytoplasmic domain. The two cytoplasmic regions bind ATP and form the catalytic core of the protein. Adenylyl cyclases are important effectors of transmembrane signaling pathways and are regulated by the activity of G protein coupled receptor signaling. This protein belongs to a small subclass of adenylyl cyclase proteins that are functionally related and are inhibited by protein kinase A, calcium ions and nitric oxide. A mutation in this gene is associated with arthrogryposis multiplex congenita.Specifications
ADCY6 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
ADCY6 | |
AC6; ACVI; Adcy6; ADCYB; adenylate cyclase 6; adenylate cyclase type 6; Adenylate cyclase type VI; adenylyl cyclase 6; ATP pyrophosphate-lyase 6; ca(2+)-inhibitable adenylyl cyclase; DKFZp779F075; KIAA0422; LCCS8; mKIAA0422 | |
ADCY6 | |
Primary | |
Antigen affinity chromatography, Protein A |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O43306, Q01341 | |
112, 11512 | |
Recombinant protein Adenylate Cyclase 6. The antigen corresponds to amino acid range 314-501 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title