Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adeno-associated virus 9 Mouse anti-Adenovirus, Unconjugated, Clone: HL2370, MilliporeSigma™
Mouse Monoclonal Antibody
$184.08 - $461.76
Specifications
Antigen | Adeno-associated virus 9 |
---|---|
Clone | HL2370 |
Applications | Dot Blot, ELISA, Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF23252MI
|
MilliporeSigma
MABF232525UL |
25 μL |
Each for $184.08
|
|
|||||
MABF23251MI
|
MilliporeSigma
MABF2325100UL |
100 μL |
Each for $461.76
|
|
|||||
Description
Adeno-associated virus 9 Monoclonal antibody specifically detects Adeno-associated virus 9 in Adenovirus samples. It is validated for Dot Blot, ELISA, Neutralizing.Specifications
Adeno-associated virus 9 | |
Dot Blot, ELISA, Neutralization | |
Unconjugated | |
Mouse | |
Adenovirus | |
Recombinant Adeno-associated virus serotype 9 capsid protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. |
HL2370 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
Adeno-associated virus serotype 9;AAV9 | |
IgG2a κ | |
Protein G Purified | |
Clone HL2370 is a mouse monoclonal antibody that specifically detects AAV9 capsid protein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title