Learn More
Thermo Scientific Chemicals Adenosine-3',5'-cyclic monophosphate sodium salt, 99%
Description
Adenosine 3?,5?-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.
Solubility
Soluble in water (50 mg/ml).
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.

Specifications
Specifications
Color | White |
Quantity | 5 g |
Beilstein | 54612 |
Solubility Information | Soluble in water (50mg/ml). |
Formula Weight | 351.19 |
Percent Purity | 99% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Adenosine-3',5'-cyclic monophosphate sodium salt |
Safety and Handling
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.